Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053124.2 | NP_444354.2 | 16 | 2456 | ||
GenBank mRNA | AF325921.1 | - | 16 | 2108 | ||
AF375046.1 | - | 16 | 2175 | |||
AK039811.1 | - | 6 | 777 | |||
AK052320.1 | - | 4 | 409 | |||
AK153789.1 | - | 16 | 2774 | |||
AK157142.1 | - | 2467 | ||||
BC021922.1 | - | 4 | 394 | |||
BC053069.1 | - | 16 | 2456 | 6409140 |
Gene Symbol: Smarca5-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079035.1 | - | 1 | 1231 |