Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033572.2 | NP_291050.1 | 8 | 1045 | ||
GenBank mRNA | AF410456.1 | - | 8 | 1024 | ||
AK005040.1 | - | 8 | 991 | |||
AK017733.1 | - | 8 | 1044 | |||
AK184744.1 | - | 2 | 105 | |||
AK185506.1 | - | 2 | 105 | |||
AK206545.1 | - | 2 | 64 | |||
BC024714.1 | - | 8 | 1017 | 5134834 | ||
BC055335.1 | - | 8 | 1043 | 6412407 |