Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP2B2 SIRNA Assays ›
Gene Symbol:
Atp2b2
Gene Name:
ATPase, Ca++ transporting, plasma membrane 2
Gene Aliases:
PMCA2
Chromosome Location:
Chr.4: 145704779 - 145948997 on Build Rnor_6.0
UniGene ID:
Rn.90982
Species:
Rat
siRNA ID
RSS302024
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Atp2b2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012508.5
NP_036640.1
976
GenBank mRNA
J03754.1
-
982
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
neural retina development
calcium ion transport
cellular calcium ion homeostasis
organelle organization
brain development
lactation
sensory perception of sound
locomotory behavior
metabolic process
regulation of cell size
cerebellum development
cerebellar Purkinje cell layer morphogenesis
cerebellar Purkinje cell differentiation
cerebellar granule cell differentiation
neuron differentiation
locomotion
serotonin metabolic process
inner ear morphogenesis
otolith mineralization
cGMP metabolic process
regulation of synaptic plasticity
inner ear development
synapse organization
neuromuscular process controlling balance
detection of mechanical stimulus involved in sensory perception of sound
regulation of cytosolic calcium ion concentration
positive regulation of calcium ion transport
auditory receptor cell stereocilium organization
inner ear receptor cell differentiation
calcium ion transmembrane transport
cochlea development
calcium ion export
calcium-transporting ATPase activity
calcium ion binding
calmodulin binding
ATP binding
protein C-terminus binding
PDZ domain binding
calcium-dependent ATPase activity
acetylcholine receptor binding
glutamate receptor binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0