Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172305.1 | NP_001165776.1 | 5, 6 | 525 | ||
NM_012713.3 | NP_036845.3 | 5, 6 | 525 | |||
GenBank mRNA | K03485.1 | - | 2, 3 | 159 | ||
M13706.1 | - | 3, 4 | 535 | |||
M19007.1 | - | 3, 4 | 485 | |||
X04440.1 | - | 3, 4 | 1079 |