Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044224.1 | NP_001037689.1 | 17 | 2861 | ||
GenBank mRNA | AB231661.1 | - | 17 | 2861 | ||
AF111111.1 | - | 5 | 433 | |||
BC166408.1 | - | 17 | 2851 | 9030944 |