Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013041.1 | NP_001013059.1 | 10, 11 | 1162 | ||
XM_006246234.2 | XP_006246296.1 | 11, 12 | 1372 | |||
XM_006246236.1 | XP_006246298.1 | 14, 15 | 1575 | |||
XM_006246237.1 | XP_006246299.1 | 12, 13 | 1277 | |||
XM_006246238.2 | XP_006246300.1 | 11, 12 | 2792 | |||
XM_008767670.1 | XP_008765892.1 | 12, 13 | 2246 | |||
XM_008767671.1 | XP_008765893.1 | 11, 12 | 2168 | |||
XM_008767672.1 | XP_008765894.1 | 11, 12 | 1211 | |||
XM_008767673.1 | XP_008765895.1 | 10, 11 | 2864 | |||
XM_008767674.1 | XP_008765896.1 | 9, 10 | 2651 | |||
XM_008767675.1 | XP_008765897.1 | 12, 13 | 1438 | |||
XM_008767676.1 | XP_008765898.1 | 10, 11 | 1163 | |||
GenBank mRNA | BC079006.1 | - | 10, 11 | 1162 | 7113075 | |
FQ223275.1 | - | 13, 14 | 3222 |