Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103689975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751137.3 | XP_003751185.2 | 4, 5 | 1013 | ||
XM_006248988.2 | XP_006249050.1 | 4, 5 | 1013 |
Gene Symbol: Ints1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769039.1 | XP_008767261.1 | 4, 5 | 1014 |