Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025403.1 | NP_001020574.1 | 2, 3 | 237 | ||
XM_006240742.2 | XP_006240804.1 | 5, 6 | 392 | |||
XM_006240744.2 | XP_006240806.1 | 3, 4 | 561 | |||
XM_008765021.1 | XP_008763243.1 | 4, 5 | 648 | |||
XM_008765022.1 | XP_008763244.1 | 4, 5 | 646 | |||
XM_008765023.1 | XP_008763245.1 | 2, 3 | 385 | |||
GenBank mRNA | BC097377.1 | - | 2, 3 | 237 | 7445017 |