Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006968.1 | NP_001006969.1 | 844 | |||
XM_006228017.2 | XP_006228079.1 | 1278 | ||||
XM_006228018.2 | XP_006228080.1 | 852 | ||||
GenBank mRNA | BC083767.1 | - | 844 | 7193201 |
Gene Symbol: LOC100909510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748716.3 | XP_003748764.1 | 4, 5 | 1214 | ||
XM_003748717.3 | XP_003748765.1 | 4, 5 | 852 | |||
XM_006228075.2 | XP_006228137.1 | 4, 5 | 1278 |