Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106578.1 | NP_001100048.1 | 15, 16 | 1715 | ||
XM_008762646.1 | XP_008760868.1 | 16, 17 | 2022 | |||
GenBank mRNA | BC169108.1 | - | 15, 16 | 1715 | 7096896 |