Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106578.1 | NP_001100048.1 | 5 | 533 | ||
XM_008762645.1 | XP_008760867.1 | 6 | 842 | |||
XM_008762646.1 | XP_008760868.1 | 6 | 840 | |||
GenBank mRNA | BC169108.1 | - | 5 | 533 | 7096896 |