Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764109.1 | XP_008762331.1 | 8 | 1113 | ||
XM_008764110.1 | XP_008762332.1 | 8 | 1113 | |||
XM_008764111.1 | XP_008762333.1 | 8 | 838 |