Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106807.1 | NP_001100277.1 | 5 | 1314 | ||
XM_006242635.2 | XP_006242697.1 | 5 | 1635 |