Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013117.2 | NP_001013135.2 | 9 | 1880 | ||
XM_006246960.2 | XP_006247022.1 | 8 | 1966 | |||
XM_008768005.1 | XP_008766227.1 | 8 | 1966 |