Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013991.1 | NP_001014013.1 | 2 | 189 | ||
XM_006250558.2 | XP_006250620.1 | 2 | 645 | |||
XM_008769936.1 | XP_008768158.1 | 1 | 1037 | |||
GenBank mRNA | BC079245.1 | - | 2 | 189 | 7132812 |