Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080149.1 | NP_001073618.1 | 8 | 1066 | ||
XM_006252765.2 | XP_006252827.1 | 9 | 1139 | |||
XM_006252767.2 | XP_006252829.1 | 9 | 1106 | |||
XM_008771099.1 | XP_008769321.1 | 9 | 1092 | |||
XM_008771100.1 | XP_008769322.1 | 9 | 1139 | |||
GenBank mRNA | BC129117.1 | - | 8 | 1066 | 7114504 |