Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108250.1 | NP_001101720.1 | 13 | 1452 | ||
XM_006256731.2 | XP_006256793.1 | 12 | 1515 | |||
GenBank mRNA | AB232988.1 | - | 13 | 1476 | ||
AB232989.1 | - | 12 | 1394 | |||
AB232990.1 | - | 12 | 1399 |