Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277386.1 | NP_001264315.1 | 876 | |||
NM_001277425.1 | NP_001264354.1 | 1641 |