Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047906.2 | NP_001041371.2 | 2 | 215 | ||
GenBank mRNA | AY325173.1 | - | 289 | |||
BC099199.1 | - | 2 | 189 | 7383928 | ||
DY575212.1 | - | 2 | 215 | |||
FQ225106.1 | - | 2 | 195 |