Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191897.1 | NP_001178826.1 | 2, 3 | 231 | ||
XM_006231063.2 | XP_006231125.1 | 2, 3 | 1723 | |||
XM_008760241.1 | XP_008758463.1 | 2, 3 | 1737 |