Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022276.1 | NP_071612.1 | 1 | 1413 | ||
XM_006231147.2 | XP_006231209.1 | 3 | 1022 | |||
XM_006231150.2 | XP_006231212.1 | 3 | 1114 | |||
GenBank mRNA | S79797.1 | - | 1 | 1413 |