Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031710.1 | NP_113898.1 | 1 | 757 | ||
GenBank mRNA | M64386.1 | - | 1 | 757 |
Gene Symbol: LOC100911376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229962.1 | XP_006230024.1 | 1 | 757 |