Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053562.3 | NP_446014.2 | 13 | 1430 | ||
GenBank mRNA | AF035673.1 | - | 13 | 1432 |