Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508795.1 | - | 2 | 183 | ||
BC062403.1 | - | 2 | 155 | 5621394 | ||
CK480694.1 | - | 2 | 148 | |||
FQ209590.1 | - | 2 | 166 | |||
FQ218571.1 | - | 2 | 168 | |||
FQ220674.1 | - | 2 | 166 | |||
M15327.1 | - | 2 | 139 |