Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007637.1 | NP_001007638.1 | 3 | 315 | ||
XM_006232604.2 | XP_006232666.1 | 3 | 607 | |||
XM_006232605.2 | XP_006232667.1 | 3 | 610 | |||
XM_006232607.2 | XP_006232669.1 | 3 | 497 | |||
XM_006232608.2 | XP_006232670.1 | 3 | 527 | |||
GenBank mRNA | BC082018.1 | - | 3 | 315 | 7132470 |