Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107462.1 | NP_001100932.1 | 29 | 3812 | ||
XM_006253759.2 | XP_006253821.1 | 33 | 4098 | |||
XM_006253760.2 | XP_006253822.1 | 33 | 4098 | |||
XM_006253762.2 | XP_006253824.1 | 33 | 4098 | |||
XM_006253763.2 | XP_006253825.1 | 32 | 3978 | |||
XM_006253764.2 | XP_006253826.1 | 32 | 3978 | |||
XM_006253765.2 | XP_006253827.1 | 31 | 3939 | |||
XM_006253766.1 | XP_006253828.1 | 29 | 3806 |