Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184051.1 | NP_908940.1 | 3 | 272 | ||
XM_006235949.1 | XP_006236011.1 | 7 | 940 | |||
XM_006235950.1 | XP_006236012.1 | 7 | 937 | |||
XM_006235951.1 | XP_006236013.1 | 7 | 940 | |||
XM_006235952.1 | XP_006236014.1 | 7 | 1843 | |||
XM_006235953.2 | XP_006236015.1 | 7 | 3549 | |||
XM_006235955.2 | XP_006236017.1 | 5 | 1813 | |||
XM_006235957.2 | XP_006236019.1 | 3 | 4576 | |||
XM_008762653.1 | XP_008760875.1 | 4 | 824 | |||
XM_008762654.1 | XP_008760876.1 | 3 | 4572 | |||
GenBank mRNA | AY348865.1 | - | 3 | 272 | ||
AY376854.1 | - | 2 | 125 | |||
BC097267.1 | - | 5 | 519 | |||
FQ223364.1 | - | 7 | 1530 |