Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022528.2 | NP_071973.1 | 4 | 376 | ||
GenBank mRNA | AJ277827.1 | - | 4 | 377 |