Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171996.3 | NP_741993.1 | 1 | 119 | ||
XM_006246024.2 | XP_006246086.1 | 2 | 319 | |||
GenBank mRNA | AF044574.1 | - | 1 | 41 | ||
BC070959.1 | - | 1 | 120 | 7097883 | ||
FQ218511.1 | - | 1 | 94 |