Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amy1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010970.1 | NP_001010970.1 | 10, 11 | 1427 | ||
GenBank mRNA | AB057450.1 | - | 10, 11 | 1427 | ||
BC088228.1 | - | 9, 10 | 1370 | 7323054 | ||
FQ219213.1 | - | 10, 11 | 1756 |