Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013076.3 | NP_037208.1 | 2 | 305 | ||
XM_008762762.1 | XP_008760984.1 | 2 | 393 | |||
GenBank mRNA | D45862.1 | - | 3 | 317 | ||
D49653.1 | - | 2 | 286 | |||
S78586.1 | - | 2 | 257 | |||
U48849.1 | - | 2 | 218 |