Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013222.2 | NP_037354.2 | 2 | 443 | ||
GenBank mRNA | D30735.1 | - | 2 | 443 |
Gene Symbol: LOC100912596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752199.3 | XP_003752247.2 | 2 | 1657 |