Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1311558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760532.1 | XP_008758754.1 | 6 | 614 | ||
XM_008760533.1 | XP_008758755.1 | 6 | 631 | |||
GenBank mRNA | BC085922.1 | - | 8 | 777 | ||
CO392889.1 | - | 490 | ||||
EF055485.1 | - | 9 | 787 | |||
EF055488.1 | - | 9 | 787 |