Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106249.2 | NP_001099719.2 | 2 | 137 | ||
XM_006228946.2 | XP_006229008.1 | 2 | 250 | |||
XM_008759327.1 | XP_008757549.1 | 2 | 225 | |||
XM_008759328.1 | XP_008757550.1 | 2 | 225 | |||
XM_008759329.1 | XP_008757551.1 | 2 | 227 |