Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017349.2 | NP_059045.2 | 2 | 78 | ||
NM_145717.1 | NP_663769.1 | 3 | 122 | |||
XM_006242911.2 | XP_006242973.1 | 3 | 279 | |||
XM_006242912.2 | XP_006242974.1 | 3 | 227 | |||
GenBank mRNA | AF129400.1 | - | 3 | 122 | ||
AF233060.1 | - | 2 | 78 | |||
X70062.1 | - | 3 | 86 |