Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024872.1 | NP_001020043.1 | 2, 3 | 551 | ||
GenBank mRNA | BC098011.1 | - | 2, 3 | 551 | 7454308 | |
FQ234084.1 | - | 2, 3 | 587 |