Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024872.1 | NP_001020043.1 | 7 | 1006 | ||
GenBank mRNA | BC098011.1 | - | 7 | 1006 | 7454308 |