Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017459.2 | NP_001017459.2 | 6 | 981 | ||
XM_006241385.2 | XP_006241447.1 | 6 | 1018 | |||
XM_006241387.2 | XP_006241449.1 | 6 | 1005 | |||
XM_006241388.2 | XP_006241450.1 | 6 | 1018 | |||
XM_008765400.1 | XP_008763622.1 | 8 | 1295 | |||
GenBank mRNA | BC087101.1 | - | 6 | 947 | 7192579 |