Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF14 SIRNA Assays ›
Gene Symbol:
Kif14
Gene Name:
kinesin family member 14
Gene Aliases:
-
Chromosome Location:
Chr.13: 53350073 - 53421992 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
siRNA ID
RSS359896
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Kif14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_008769585.1
XP_008767807.1
4, 5
1810
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
microtubule-based movement
microtubule depolymerization
mitotic metaphase plate congression
metabolic process
positive regulation of cell proliferation
regulation of G2/M transition of mitotic cell cycle
cerebellar granular layer structural organization
cerebellar Purkinje cell layer structural organization
cerebellar cortex development
hippocampus development
olfactory bulb development
cell proliferation in forebrain
cerebral cortex development
regulation of cell adhesion
regulation of cell migration
cytoskeleton-dependent intracellular transport
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
regulation of myelination
activation of protein kinase activity
positive regulation of cytokinesis
regulation of Rap protein signal transduction
negative regulation of integrin activation
substrate adhesion-dependent cell spreading
negative regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
establishment of protein localization
cell division
response to docetaxel trihydrate
mitotic cell cycle process
regulation of cell maturation
regulation of G1/S transition of mitotic cell cycle
microtubule motor activity
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
tubulin binding
ATPase activity
protein kinase binding
PDZ domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0