Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013216.2 | NP_001013234.2 | 7 | 1434 | ||
XM_006239469.2 | XP_006239531.1 | 7 | 1443 | |||
XM_008764367.1 | XP_008762589.1 | 7 | 1472 | |||
GenBank mRNA | BC091214.1 | - | 6 | 2033 | 7383542 | |
FQ211844.1 | - | 7 | 1461 |