Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssuh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729443.4 | XP_002729489.2 | 5 | 443 | ||
XM_006237061.2 | XP_006237123.1 | 5 | 440 |