Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 141 |
Gene Symbol: LOC100360061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729626.3 | XP_002729672.1 | 123 |
Gene Symbol: LOC100909786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750168.3 | XP_003750216.1 | 1 | 123 |
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233682.2 | XP_006233744.1 | 1, 2 | 136 |