Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109328.2 | NP_001102798.1 | 4 | 327 | ||
XM_008769490.1 | XP_008767712.1 | 4 | 396 | |||
XM_008769491.1 | XP_008767713.1 | 4 | 742 | |||
XM_008769492.1 | XP_008767714.1 | 4 | 743 | |||
XM_008769493.1 | XP_008767715.1 | 4 | 747 | |||
GenBank mRNA | FM076322.1 | - | 4 | 327 |