Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099482.1 | NP_001092952.1 | 6 | 2529 |
Gene Symbol: LOC103689970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765901.1 | XP_008764123.1 | 4 | 1287 |
Set Membership: |
G-protein coupled receptor |