Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC678893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750190.3 | XP_003750238.1 | 3 | 677 | ||
XM_008764871.1 | XP_008763093.1 | 1 | 2604 | |||
XM_008764872.1 | XP_008763094.1 | 3 | 2874 | |||
XM_008764873.1 | XP_008763095.1 | 4 | 3399 | |||
XM_008764874.1 | XP_008763096.1 | 4 | 1064 | |||
XM_008764875.1 | XP_008763097.1 | 3 | 940 | |||
XM_008764876.1 | XP_008763098.1 | 3 | 1186 | |||
XM_008764877.1 | XP_008763099.1 | 4 | 2937 |