Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053873.1 | NP_446325.1 | 2074 | |||
GenBank mRNA | AB374360.1 | - | 2074 |