Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002847.2 | 1 | 5529 | |||
GenBank mRNA | AK141413.1 | - | 1 | 612 | ||
EF177380.1 | - | 1 | 5529 | |||
FJ209304.1 | - | 1 | 5529 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AY722410.1 | - | 1 | 5529 | Chr. - - [-] on Build |