Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa11os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015348.1 | 1 | 437 | |||
GenBank mRNA | U20366.1 | - | LIT1565 Hoxa11as |
1 | 437 |