Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024509.1 | 3 | 616 | |||
GenBank mRNA | AK148367.1 | - | 3 | 587 | ||
BC096570.1 | - | 3 | 570 |