Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5830417I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028359.1 | 18 | 6165 | |||
GenBank mRNA | AK017936.1 | - | 4 | 1583 | ||
AK049055.1 | - | 3 | 607 | |||
BC040255.1 | - | 3610 | ||||
BC059914.1 | - | 3 | 3289 |
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY040842.1 | - | 3827 |